Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lztfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033322.2 | NP_201579.1 | 8 | 695 | ||
GenBank mRNA | AJ297352.1 | - | 8 | 691 | ||
AK029671.1 | - | 8 | 727 | |||
AK030068.1 | - | 8 | 707 | |||
AK041091.1 | - | 8 | 728 | |||
AK046515.1 | - | 8 | 684 | |||
AK054003.1 | - | 8 | 701 | |||
AK081529.1 | - | 6 | 480 | |||
AK145263.1 | - | 8 | 704 | |||
AK147312.1 | - | 7 | 639 | |||
AK157770.1 | - | 8 | 702 | |||
BC026840.1 | - | 8 | 695 | 4165152 | ||
BC037045.1 | - | 8 | 695 | |||
BC098224.1 | - | 8 | 700 | 1263280 |