Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164359.1 | NP_001157831.1 | 3 | 353 | ||
NM_001164360.1 | NP_001157832.1 | 3 | 261 | |||
NM_145502.3 | NP_663477.3 | 2 | 457 | |||
GenBank mRNA | AK133557.1 | - | 2 | 200 | ||
AK145990.1 | - | 3 | 234 | |||
AK171750.1 | - | 1 | 1746 | |||
BC011220.1 | - | 3 | 225 | 4208238 | ||
BC021793.1 | - | 2 | 515 | |||
BC023849.1 | - | 2 | 320 | 5345801 | ||
CJ074662.1 | - | 3 | 262 | |||
CJ134812.1 | - | 3 | 334 | |||
CN702299.1 | - | 2 | 457 |