Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081304.1 | NP_001074773.1 | 1, 2 | 131 | ||
GenBank mRNA | AK020270.1 | - | 1, 2 | 111 | ||
AK036106.1 | - | 1, 2 | 111 | |||
AK036335.1 | - | 1, 2 | 129 | |||
AK087000.1 | - | 1, 2 | 112 |