Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110791.1 | NP_001104261.1 | 9 | 1315 | ||
NM_011750.2 | NP_035880.2 | 9 | 1315 | |||
GenBank mRNA | AK133895.1 | - | 9 | 1104 | ||
AK146151.1 | - | 9 | 1076 | |||
AK147081.1 | - | 9 | 1084 | |||
AK152914.1 | - | 8 | 1018 | |||
AK154705.1 | - | 9 | 1083 | |||
AK157845.1 | - | 9 | 1084 | |||
AK195027.1 | - | 3 | 214 | |||
BC009091.1 | - | 9 | 1060 | 3157495 | ||
BC055370.1 | - | 9 | 1074 | 30055109 | ||
CD563870.1 | - | 3 | 176 | |||
X80159.1 | - | 1 | 32 | |||
X85802.1 | - | 9 | 1072 |