Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011753.3 | NP_035883.2 | 7, 8 | 898 | ||
GenBank mRNA | AK029298.1 | - | 5, 6 | 1163 | ||
AK082002.1 | - | 7, 8 | 732 | |||
AK141740.1 | - | 6, 7 | 1261 | |||
U29504.1 | - | 47 |