Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011753.3 | NP_035883.2 | 10 | 3213 | ||
GenBank mRNA | AK141740.1 | - | 9 | 3576 | ||
AK220539.1 | - | 1 | 1190 | |||
M36514.1 | - | 1 | 1165 | |||
X12592.1 | - | 1 | 1165 |