Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146107.2 | NP_666219.1 | 7 | 787 | ||
GenBank mRNA | AK078508.1 | - | 7 | 803 | ||
AK088839.1 | - | 7 | 820 | |||
AK178242.1 | - | 1 | 73 | |||
AK205839.1 | - | 2 | 154 | |||
BC023020.1 | - | 7 | 787 | 5375425 | ||
BC024861.1 | - | 2 | 111 |