Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160038.1 | NP_001153510.1 | 15 | 2033 | ||
NM_001160039.1 | NP_001153511.1 | 15 | 1875 | |||
NM_001160040.1 | NP_001153512.1 | 14 | 1929 | |||
NM_145518.2 | NP_663493.2 | 14 | 1771 | |||
GenBank mRNA | AK034597.1 | - | 14 | 1840 | ||
AK036926.1 | - | 14 | 1636 | |||
AK134505.1 | - | 1 | 1269 | |||
AK142711.1 | - | 14 | 1651 | |||
AK146814.1 | - | 14 | 1789 | |||
AK167705.1 | - | 14 | 1619 | |||
AK196215.1 | - | 1 | 18 | |||
AK219412.1 | - | 1 | 18 | |||
BC006660.1 | - | 14 | 1658 | 3501043 | ||
BC015300.1 | - | 14 | 1591 | 4236882 |