Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160038.1 | NP_001153510.1 | 9 | 1203 | ||
NM_001160039.1 | NP_001153511.1 | 9 | 1045 | |||
NM_001160040.1 | NP_001153512.1 | 8 | 1099 | |||
NM_145518.2 | NP_663493.2 | 8 | 941 | |||
GenBank mRNA | AK034597.1 | - | 8 | 1010 | ||
AK036926.1 | - | 8 | 806 | |||
AK078590.1 | - | 8 | 788 | |||
AK142711.1 | - | 8 | 821 | |||
AK146814.1 | - | 8 | 959 | |||
AK166604.1 | - | 9 | 892 | |||
AK167705.1 | - | 8 | 789 | |||
BC006660.1 | - | 8 | 828 | 3501043 | ||
BC015300.1 | - | 8 | 761 | 4236882 |