Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppip5k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173760.5 | NP_776121.4 | 19, 20 | 2811 | ||
GenBank mRNA | AK087712.1 | - | 19, 20 | 2741 | ||
AK129140.1 | - | 19, 20 | 2747 | |||
AK138622.1 | - | 19, 20 | 3128 | |||
AK149804.1 | - | 19, 20 | 2743 | |||
AK162179.1 | - | 12, 13 | 1530 | |||
BC046411.1 | - | 9, 10 | 1124 | |||
BC053396.1 | - | 18, 19 | 2313 | 30044788 |