Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppip5k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173760.5 | NP_776121.4 | 8 | 1395 | ||
GenBank mRNA | AK087712.1 | - | 8 | 1325 | ||
AK129140.1 | - | 8 | 1331 | |||
AK138622.1 | - | 8 | 1715 | |||
AK149804.1 | - | 8 | 1327 | |||
AK162179.1 | - | 1 | 112 | |||
AK166095.1 | - | 8 | 1396 | |||
BC053396.1 | - | 7 | 897 | 30044788 |