Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290374.1 | NP_001277303.1 | 4 | 974 | ||
NM_001290375.1 | NP_001277304.1 | 5 | 1049 | |||
NM_001290376.1 | NP_001277305.1 | 5 | 565 | |||
NM_177343.4 | NP_796317.2 | 5 | 1049 | |||
GenBank mRNA | AK052401.1 | - | 5 | 470 | ||
AK053173.1 | - | 5 | 803 | |||
AK147616.1 | - | 5 | 1050 | |||
AK147657.1 | - | 5 | 760 | |||
AK154434.1 | - | 5 | 638 | |||
AK162754.1 | - | 5 | 764 | |||
AK170123.1 | - | 4 | 733 | |||
AK170777.1 | - | 5 | 814 | |||
AY273822.1 | - | 5 | 565 | |||
BC141413.1 | - | 5 | 549 | 9056026 | ||
BC141414.1 | - | 5 | 549 | 9056027 |