Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009584.4 | NP_033610.1 | 13 | 1444 | ||
GenBank mRNA | AK131964.1 | - | 13 | 1465 | ||
AK167991.1 | - | 13 | 1465 | |||
BC021755.1 | - | 12 | 3189 | |||
BC052027.1 | - | 13 | 1471 | 5718637 | ||
D63784.1 | - | 13 | 1443 | |||
U53208.1 | - | 13 | 1399 |