Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145832.3 | NP_665831.1 | 2 | 260 | ||
NM_211358.2 | NP_997597.1 | 1 | 748 | |||
GenBank mRNA | AJ440720.1 | - | 1 | 145 | ||
AK030977.1 | - | 2 | 261 | |||
AK040986.1 | - | 1 | 495 | |||
AK045972.1 | - | 2 | 237 | |||
AK050311.1 | - | 2 | 225 | |||
AK081068.1 | - | 1 | 246 | |||
AK088227.1 | - | 2 | 247 | |||
AK138595.1 | - | 1 | 691 | |||
AK144762.1 | - | 1 | 670 | |||
AK151154.1 | - | 1 | 744 | |||
AK181528.1 | - | 1 | 47 | |||
BC031814.1 | - | 1 | 342 | 4196818 | ||
BC116736.1 | - | 1 | 173 | 40126055 | ||
BC119128.1 | - | 1 | 173 | 8733877 | ||
BY026060.1 | - | 2 | 261 |