Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033296.2 | NP_001028468.1 | 20 | 2118 | ||
GenBank mRNA | AK016809.1 | - | 20 | 2064 | ||
AK132650.1 | - | 20 | 2119 | |||
BC150948.1 | - | 20 | 2090 | 9087859 |