Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031363.1 | - | 1, 2 | 292 | ||
AK032644.1 | - | 1, 2 | 263 | |||
AK045891.1 | - | 1, 2 | 275 | |||
AK053924.1 | - | 1, 2 | 293 | |||
AK078867.1 | - | 1, 2 | 251 | |||
BC012260.1 | - | 1, 2 | 254 | |||
BC128314.1 | - | 37 | ||||
BC131928.1 | - | 1, 2 | 156 | 40130205 | ||
BC131930.1 | - | 1, 2 | 156 | 40130207 | ||
BE649179.1 | - | 1, 2 | 307 |