Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144500.3 | NP_653083.2 | 9 | 1015 | ||
GenBank mRNA | AK048537.1 | - | 9 | 935 | ||
AK145292.1 | - | 9 | 1047 | |||
AK161695.1 | - | 9 | 954 | |||
AK166059.1 | - | 9 | 994 | |||
AK169504.1 | - | 2 | 940 | |||
AK170306.1 | - | 9 | 999 | |||
AK170669.1 | - | 9 | 1016 | |||
AK178638.1 | - | 3 | 194 | |||
AK182769.1 | - | 3 | 203 | |||
AK220257.1 | - | 3 | 334 | |||
BC026804.1 | - | 9 | 1022 | 4159217 |