Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm4sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145539.2 | NP_663514.2 | 1 | 292 | ||
GenBank mRNA | AK148484.1 | - | 1 | 260 | ||
AK165731.1 | - | 1 | 260 | |||
BB840196.1 | - | 1 | 389 | |||
BC010814.1 | - | 1 | 243 | 4211816 |