Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178246.3 | NP_839977.2 | 8 | 923 | ||
GenBank mRNA | AK129286.1 | - | 8 | 902 | ||
AK143911.1 | - | 8 | 925 | |||
AK155787.1 | - | 8 | 912 | |||
AK172217.1 | - | 7 | 803 | |||
BC024683.1 | - | 8 | 875 | 3992000 | ||
BC100408.1 | - | 8 | 872 | 30734268 |