Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatad2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139304.1 | NP_647465.1 | 5 | 795 | ||
GenBank mRNA | AF411837.1 | - | 5 | 795 | ||
AK041594.1 | - | 6 | 852 | |||
AK141586.1 | - | 7 | 1082 | |||
AK173113.1 | - | 2 | 113 | |||
BC130262.1 | - | 5 | 797 | 40142858 |