Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080818.2 | NP_001074287.1 | 8 | 1123 | ||
NM_001173553.1 | NP_001167024.1 | 6 | 976 | |||
GenBank mRNA | AK036556.1 | - | 5 | 364 | ||
AK043956.1 | - | 8 | 1108 | |||
AK048250.1 | - | 8 | 1028 | |||
AK052560.1 | - | 8 | 1114 | |||
AK142294.1 | - | 8 | 1123 | |||
BC072644.1 | - | 6 | 930 | 30356962 |