Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173762.4 | NP_776123.3 | 22 | 2399 | ||
GenBank mRNA | AY493378.1 | - | 22 | 2303 |