Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znhit6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081094.1 | NP_001074563.1 | 1 | 386 | ||
GenBank mRNA | AK148729.1 | - | 1 | 363 | ||
BC147498.1 | - | 1 | 272 | 9056360 | ||
BC150966.1 | - | 1 | 272 | 9087878 | ||
BC150967.1 | - | 1 | 272 | 9087879 | ||
BC151198.1 | - | 1 | 272 | 9088086 |