Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290645.1 | NP_001277574.1 | 6, 7 | 1051 | ||
NM_146148.2 | NP_666260.1 | 7, 8 | 1148 | |||
GenBank mRNA | AB077295.1 | - | 7, 8 | 1162 | ||
AB077296.1 | - | 7, 8 | 1111 | |||
AB077297.1 | - | 6, 7 | 1017 | |||
AB077298.1 | - | 7, 8 | 1111 | |||
AK090245.1 | - | 7, 8 | 1148 | |||
BC027748.1 | - | 7, 8 | 1144 | 5052412 |