Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013756.1 | NP_001013778.1 | 1, 2 | 202 | ||
GenBank mRNA | BC089372.1 | - | 1, 2 | 202 | 30607679 |