Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039200.2 | NP_001034289.1 | 15 | 1716 | ||
NM_146157.4 | NP_666269.2 | 16 | 1725 | |||
GenBank mRNA | AF525925.1 | - | 6 | 566 | ||
AK049061.1 | - | 16 | 1716 | |||
AK077496.1 | - | 16 | 1716 | |||
AK089289.1 | - | 15 | 1695 | |||
AK129052.1 | - | 16 | 1678 | |||
AK138637.1 | - | 16 | 1698 | |||
AK161677.1 | - | 15 | 1679 | |||
AK170739.1 | - | 15 | 1695 | |||
BC048948.1 | - | 2 | 271 | |||
BC056952.1 | - | 11 | 1445 | |||
BC129804.1 | - | 16 | 1712 | 40087024 | ||
BC139366.1 | - | 16 | 1711 | 8862388 |