Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112744.1 | NP_001106215.1 | 11 | 1679 | ||
GenBank mRNA | AK042195.1 | - | 11 | 1675 | ||
AK153708.1 | - | 11 | 1840 | |||
BC020030.1 | - | 8 | 840 | |||
BC026661.1 | - | 3 | 692 | |||
BC044805.1 | - | 10 | 3093 | |||
BC095966.1 | - | 9 | 976 | |||
BC137847.1 | - | 11 | 1720 | 8860866 | ||
BC144970.1 | - | 11 | 1720 | 9053518 | ||
BC144971.1 | - | 11 | 1743 |