Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aasdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173765.3 | NP_776126.1 | 10, 11 | 1885 | ||
GenBank mRNA | AB095954.1 | - | 10, 11 | 1902 | ||
AK038779.1 | - | 11, 12 | 1895 | |||
AK136849.1 | - | 10, 11 | 1883 | |||
AK148405.1 | - | 9, 10 | 1711 | |||
AK215879.1 | - | 1, 2 | 169 | |||
AK218868.1 | - | 1, 2 | 169 | |||
BC058102.1 | - | 10, 11 | 1870 | |||
BC128330.1 | - | 9, 10 | 1757 | 40110405 |