Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163461.2 | NP_001156933.1 | 8 | 1176 | ||
NM_001163462.2 | NP_001156934.1 | 8 | 1233 | |||
NM_001289569.1 | NP_001276498.1 | 8 | 1190 | |||
NM_001289570.1 | NP_001276499.1 | 8 | 1176 | |||
NM_144911.3 | NP_659160.2 | 8 | 1176 | |||
GenBank mRNA | AK034418.1 | - | 8 | 1192 | ||
AK049889.1 | - | 8 | 1233 | |||
AK145830.1 | - | 8 | 1151 | |||
AK161901.1 | - | 8 | 1157 | |||
AK166574.1 | - | 8 | 1099 | |||
BC021895.1 | - | 8 | 1137 | 3481916 |