Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aimp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172146.1 | NP_001165617.1 | 3 | 590 | ||
NM_146165.2 | NP_666277.1 | 3 | 470 | |||
GenBank mRNA | BC024410.1 | - | 3 | 563 | 4954972 | |
BC024480.1 | - | 3 | 461 | 4977193 | ||
BC026958.1 | - | 3 | 529 | 4013019 | ||
BC026972.1 | - | 3 | 543 | 5371230 | ||
CF732613.1 | - | 3 | 512 | |||
CJ051555.1 | - | 3 | 462 | |||
CX566811.1 | - | 1 | 98 |