Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178703.4 | NP_848818.1 | 4 | 641 | ||
GenBank mRNA | AK042956.1 | - | 4 | 503 | ||
AK047663.1 | - | 4 | 487 | |||
AK052971.1 | - | 4 | 466 | |||
AK053798.1 | - | 480 | ||||
AK053831.1 | - | 3 | 800 | |||
AK053883.1 | - | 4 | 463 | |||
BC059080.1 | - | 4 | 492 | 6827121 | ||
M92378.1 | - | 3 | 403 |