Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gys2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145572.2 | NP_663547.2 | 6 | 1233 | ||
GenBank mRNA | AK138992.1 | - | 6 | 941 | ||
AK149391.1 | - | 6 | 957 | |||
BC021322.1 | - | 6 | 1094 | 5051685 | ||
BC158081.1 | - | 6 | 1094 | 9007373 |