Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caprin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301351.1 | NP_001288280.1 | 18 | 2647 | ||
NM_181541.4 | NP_853519.2 | 19 | 3307 | |||
GenBank mRNA | AK129460.1 | - | 18 | 2560 | ||
AK140744.1 | - | 19 | 3310 | |||
BC025636.1 | - | 19 | 3263 | |||
BC027523.1 | - | 1 | 773 | |||
BC092093.1 | - | 5 | 929 | |||
BC125373.1 | - | 19 | 3196 | 40129888 | ||
BC125375.1 | - | 19 | 3196 | 40129890 | ||
BE946020.1 | - | 1 | 403 |