Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144922.2 | NP_659171.1 | 6 | 902 | ||
NM_178089.2 | NP_835190.1 | 6 | 883 | |||
GenBank mRNA | AK042090.1 | - | 6 | 883 | ||
AK087545.1 | - | 6 | 872 | |||
AK143666.1 | - | 6 | 945 | |||
AK155604.1 | - | 6 | 913 | |||
AK195535.1 | - | 2 | 133 | |||
BC021506.1 | - | 6 | 926 | 5347099 | ||
BC027844.1 | - | 3 | 195 | |||
BC078642.1 | - | 6 | 826 | 6329630 |