Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hipk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033315.2 | NP_001028487.2 | 3 | 1136 | ||
GenBank mRNA | AB354583.1 | - | 3 | 880 | ||
AK133498.1 | - | 2 | 1653 | |||
BC147572.1 | - | 2 | 296 | 9056562 | ||
BC147577.1 | - | 2 | 296 | 9056567 |