Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289670.1 | NP_001276599.1 | 12, 13 | 1209 | ||
NM_001289682.1 | NP_001276611.1 | 13, 14 | 1292 | |||
NM_178252.3 | NP_839983.1 | 13, 14 | 1319 | |||
GenBank mRNA | AK142180.1 | - | 12, 13 | 1210 | ||
AY217764.1 | - | 12, 13 | 1152 | |||
BC061471.1 | - | 6, 7 | 496 | 5698780 | ||
BC065086.1 | - | 13, 14 | 1268 | 6850475 | ||
BC065166.1 | - | 13, 14 | 1266 | 6836510 | ||
BC066047.1 | - | 13, 14 | 1244 | 6822191 |