Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173767.3 | NP_776128.2 | 9, 10 | 1450 | ||
GenBank mRNA | AK050143.1 | - | 6, 7 | 945 | ||
AK178410.1 | - | 1, 2 | 124 | |||
AK196619.1 | - | 1, 2 | 124 | |||
BC026405.1 | - | 9, 10 | 1337 | 4211657 | ||
BC089586.1 | - | 6, 7 | 954 | |||
BC139196.1 | - | 6, 7 | 838 | 8862218 | ||
BC139197.1 | - | 6, 7 | 838 | 8862219 | ||
DQ205645.1 | - | 8, 9 | 1163 |