Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033318.3 | NP_001028490.2 | 5, 6 | 805 | ||
GenBank mRNA | AK030709.1 | - | 5, 6 | 772 | ||
AK145163.1 | - | 5, 6 | 803 | |||
AK154693.1 | - | 5, 6 | 807 | |||
AK163143.1 | - | 825 | ||||
AK172200.1 | - | 5, 6 | 807 |