Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037736.2 | NP_001032825.1 | 18, 19 | 2374 | ||
NM_172751.3 | NP_766339.2 | 19, 20 | 2491 | |||
GenBank mRNA | AK032452.1 | - | 17, 18 | 2091 | ||
AK147669.1 | - | 19, 20 | 2492 | |||
AK220332.1 | - | 18, 19 | 2299 | |||
BC047430.1 | - | 3, 4 | 325 | |||
BC059212.1 | - | 14, 15 | 1656 | 6400425 | ||
BC066074.1 | - | 18, 19 | 2294 | 6855196 |
Gene Symbol: Uvrag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK190073.1 | - | 193 |