Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colgalt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146211.3 | NP_666323.2 | 8 | 1250 | ||
GenBank mRNA | AK029433.1 | - | 8 | 1104 | ||
AK035788.1 | - | 8 | 1128 | |||
AK036165.1 | - | 8 | 1253 | |||
AK076103.1 | - | 8 | 1106 | |||
AK154775.1 | - | 8 | 1104 | |||
AK193650.1 | - | 1 | 12 | |||
BC018287.1 | - | 8 | 1015 | |||
BC027801.1 | - | 1 | 137 | |||
BC032165.1 | - | 8 | 1282 | 5353334 | ||
BC034208.1 | - | 5 | 568 | |||
BC056951.1 | - | 8 | 1141 | 5695490 |