Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173413.3 | NP_775589.1 | 5 | 408 | ||
GenBank mRNA | AB232601.1 | - | 5 | 336 | ||
AK031251.1 | - | 5 | 355 | |||
AK051218.1 | - | 5 | 358 | |||
AK084650.1 | - | 5 | 358 | |||
AK155621.1 | - | 5 | 358 | |||
BC059208.1 | - | 5 | 390 | 6405462 | ||
BC157974.1 | - | 5 | 356 | 9007296 |