Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033329.3 | NP_001028501.1 | 5, 6 | 1051 | ||
NM_001290384.1 | NP_001277313.1 | 4, 5 | 673 | |||
NM_001290385.1 | NP_001277314.1 | 4, 5 | 1497 | |||
GenBank mRNA | AK038840.1 | - | 5, 6 | 969 | ||
AK122280.1 | - | 4, 5 | 608 | |||
AK139416.1 | - | 5, 6 | 999 | |||
AK163874.1 | - | 4, 5 | 654 | |||
BC067031.1 | - | 4, 5 | 1319 | |||
BC141385.1 | - | 4, 5 | 693 | 9055998 |