Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033329.3 | NP_001028501.1 | 9 | 1737 | ||
NM_001290384.1 | NP_001277313.1 | 8 | 1359 | |||
NM_001290385.1 | NP_001277314.1 | 8 | 2183 | |||
GenBank mRNA | AK038840.1 | - | 9 | 1655 | ||
AK122280.1 | - | 8 | 1294 | |||
AK139416.1 | - | 9 | 1685 | |||
AK163874.1 | - | 8 | 1340 | |||
BC067031.1 | - | 8 | 2005 | |||
BC141385.1 | - | 8 | 1379 | 9055998 |