Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmtc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033332.2 | NP_001028504.1 | 5 | 796 | ||
NM_001110013.1 | NP_001103483.1 | 5 | 796 | |||
GenBank mRNA | AK044857.1 | - | 5 | 752 | ||
AK149759.1 | - | 5 | 755 | |||
AK158192.1 | - | 5 | 782 | |||
AK165838.1 | - | 5 | 797 |