Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172800.2 | NP_766388.2 | 1, 2 | 154 | ||
GenBank mRNA | AK030591.1 | - | 1252 | |||
AK031283.1 | - | 1233 | ||||
AK052040.1 | - | 1, 2 | 179 | |||
AY351699.1 | - | 1, 2 | 154 | |||
BC038036.1 | - | 1, 2 | 432 |