Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177673.1 | NP_001171144.1 | 3 | 1030 | ||
NM_001177674.1 | NP_001171145.1 | 3 | 1212 | |||
NM_175493.4 | NP_780702.1 | 3 | 1333 | |||
GenBank mRNA | AK036659.1 | - | 2 | 1042 | ||
AK087410.1 | - | 3 | 1014 | |||
AK154649.1 | - | 2 | 1005 | |||
AK155886.1 | - | 2 | 1022 | |||
BC118034.1 | - | 2 | 1011 | 40106885 | ||
BC118044.1 | - | 2 | 1011 | 40106886 |
Set Membership: |
G-protein coupled receptor |