Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc24a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172152.2 | NP_742164.1 | 15 | 1546 | ||
GenBank mRNA | AK044239.1 | - | 15 | 2178 | ||
AY156046.1 | - | 15 | 1546 | |||
BC137945.1 | - | 15 | 1517 | 8860964 | ||
BC171939.1 | - | 15 | 1517 | 9054225 |