Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097621.1 | NP_001091090.1 | 12 | 3686 | ||
GenBank mRNA | BC059062.1 | - | 1 | 517 | ||
BC094358.1 | - | 1 | 1191 | |||
BM963109.1 | - | 1 | 387 | |||
GQ489027.1 | - | 12 | 3529 |