Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF086810.1 | - | 3 | 391 | ||
AF234838.1 | - | 8 | 1779 | |||
AK140319.1 | - | 8 | 1745 | |||
AK140325.1 | - | 8 | 1745 | |||
BC057334.1 | - | 8 | 1797 | 6811850 |