Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011840.2 | NP_035970.1 | 15 | 1546 | ||
GenBank mRNA | AB019374.1 | - | 15 | 1547 | ||
AK013856.1 | - | 7 | 389 | |||
AK159261.1 | - | 15 | 1596 | |||
BC028260.1 | - | 15 | 1426 | 5067818 |